missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgD Rat anti-Mouse, FITC, Clone: 11-26c (11-26), eBioscience™

Rat Monoclonal Antibody

$117.48 - $289.08

Specifications

Antigen IgD
Clone 11-26c (11-26)
Host Species Rat
Gene Alias Immunoglobulin D, Ig D
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509953 Life Technologies
LS11599381
50μg Each for $117.48
509954 Life Technologies
LS11599382
100μg Each for $196.68
509955 Life Technologies
LS11599385
500μg Each for $289.08
Specifications

Description

The 11-26c monoclonal antibody reacts with the δ heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells.
Description & Specifications

Specifications

IgD
Rat
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL
11-26c (11-26)
Immunoglobulin D, Ig D
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer