Please login to your online account to display your discounted pricing

IgD Rat anti-Mouse, eFluor 450, Clone: 11-26c (11-26), eBioscience™

Rat Monoclonal Antibody

$143.55 - $288.55

Specifications

Antigen IgD
Clone 11-26c (11-26)
Host Species Rat
Gene Alias Immunoglobulin D, Ig D
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016392

 
life technologies
LS48599382
100μg Each for $288.55

501129162

 
life technologies
LS48599380
25μg Each for $143.55
Description & Specifications

Specifications

Antigen IgD
Clone 11-26c (11-26)
Host Species Rat
Gene Alias Immunoglobulin D, Ig D
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 11-26c monoclonal antibody reacts with the δ heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells.