Please login to your online account to display your discounted pricing

IgD Rat anti-Mouse, Biotin, Clone: 11-26c (11-26), eBioscience™

Rat Monoclonal Antibody

$114.55 - $317.55

Specifications

Antigen IgD
Clone 11-26c (11-26)
Host Species Rat
Gene Alias Immunoglobulin D, Ig D
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122566

 
affymetrix
13599381
50μg Each for $114.55

501122567

 
affymetrix
13599382
100μg Each for $172.55

501128799

 
affymetrix
13599385
500μg Each for $317.55
Description & Specifications

Specifications

Antigen IgD
Clone 11-26c (11-26)
Host Species Rat
Gene Alias Immunoglobulin D, Ig D
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 11-26c monoclonal antibody reacts with the δ heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells.