Please login to your online account to display your discounted pricing

IgD Rat anti-Mouse, APC-eFluor 780, Clone: 11-26c (11-26), eBioscience™

Rat Monoclonal Antibody

$187.05 - $448.05

Specifications

Antigen IgD
Clone 11-26c (11-26)
Host Species Rat
Gene Alias Immunoglobulin D, Ig D
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124067

 
life technologies
LS47599380
25μg Each for $187.05

501124068

 
life technologies
LS47599382
100μg Each for $448.05
Description & Specifications

Specifications

Antigen IgD
Clone 11-26c (11-26)
Host Species Rat
Gene Alias Immunoglobulin D, Ig D
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC-eFluor 780
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 11-26c monoclonal antibody reacts with the delta heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells.