Please login to your online account to display your discounted pricing

IgD Mouse anti-Mouse, Clone: 10.4.22, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$114.55 - $216.05

Specifications

Antigen IgD
Clone 10.4.22
Host Species Mouse
Gene Alias Immunoglobulin D, Ig-D
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122967

 
affymetrix
16592482
100μg Each for $114.55

501128593

 
affymetrix
16592485
500μg Each for $216.05
Description & Specifications

Specifications

Antigen IgD
Clone 10.4.22
Host Species Mouse
Gene Alias Immunoglobulin D, Ig-D
Species Reactivity Mouse
Applications Functional Assay
Applications In Vivo Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

This 10.4.22 monoclonal antibody reacts with the a allotype of mouse IgD, which is expressed by mature peripheral B cells. This antibody has been reported to activate B cells via engagement of the IgD receptor. The use of anti-IgM as a polyclonal B cell activator is problematic due to interference from high levels of circulating serum IgM.

Crossblocking studies suggest that the 10.4.22 antibody recognizes a different epitope from 11-26c.