Please login to your online account to display your discounted pricing

IgD Mouse anti-Human, PE, Clone: IA6-2, eBioscience™

Mouse Monoclonal Antibody

$152.25 - $381.35

Specifications

Antigen IgD
Clone IA6-2
Host Species Murine
Gene Alias Immunoglobulin D
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122482

 
life technologies
LS12986841
25 Tests Each for $152.25

501122483

 
life technologies
LS12986842
100 Tests Each for $381.35
Description & Specifications

Specifications

Antigen IgD
Clone IA6-2
Host Species Murine
Gene Alias Immunoglobulin D
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

This IA6-2 monoclonal antibody reacts with the heavy chain of human immunoglobulin D (IgD). This type I transmembrane protein is co-expressed on the surface of mature naïve B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.