missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgD Mouse anti-Human, FITC, Clone: IA6-2, eBioscience™

Mouse Monoclonal Antibody

$133.32 - $320.76

Specifications

Antigen IgD
Clone IA6-2
Host Species Murine
Gene Alias Immunoglobulin D
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122190 Life Technologies
LS11986842
100 Tests Each for $320.76
501129069 Life Technologies
LS11986841
25 Tests Each for $133.32
Specifications

Description

This IA6-2 monoclonal antibody reacts with the heavy chain of human immunoglobulin D (IgD). This type I transmembrane protein is co-expressed on the surface of mature naïve B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.
Description & Specifications

Specifications

IgD
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
IA6-2
Immunoglobulin D
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test