missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IgA Rat anti-Mouse, FITC, Clone: mA-6E1, eBioscience™

Rat Monoclonal Antibody

$132.00 - $394.68

Specifications

Antigen IgA
Clone mA-6E1
Host Species Rat
Gene Alias Immunoglobulin A
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122116 Life Technologies
LS11420483
200μg Each for $394.68
509807 Life Technologies
LS11420481
50μg Each for $132.00
509808 Life Technologies
LS11420482
100μg Each for $262.68
Specifications

Description

The mA-6E1 monoclonal antibody reacts with the heavy chain of mouse IgA. It does not react with other isotypes of antibody, including IgD, IgG or IgM, or with other species isotypes. IgA, which is mainly found as a dimer through association of the J chain, is found in mucous secretions, but not in plasma. Fluorochrome conjugated mA-6E1 is used as a detection secondary for mouse IgA primary antibodies.
Description & Specifications

Specifications

IgA
Rat
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL
mA-6E1
Immunoglobulin A
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer