missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ IgA (α -Heavy Chain) Ab-1, Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ IgA (α-Heavy Chain) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

$239.61 - $700.73

Specifications

Antigen IgA (alpha-Heavy Chain) Ab-1
Clone GA01
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS1008P Thermo Scientific™ Lab Vision™
MS1008P
200μg/mL 1mL Each for $602.71
MS1008P0 Thermo Scientific™ Lab Vision™
MS1008P0
200μg/mL 100μL Each for $239.61
MS1008P1 Thermo Scientific™ Lab Vision™
MS1008P1
200μg/mL 500μL Each for $414.59
MS1008P1ABX Thermo Scientific
MS1008P1ABX
1mg/mL 100μL Each for $545.18
MS1008PABX Thermo Scientific™ Lab Vision™
MS1008PABX
1mg/mL 200μL Each for $700.73
Specifications

Description

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ IgA (α-Heavy Chain) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: GA01

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Third constant domain (CH3) of the α-chain of IgA

Immunogen: Purified human α-heavy chain

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

IgA (alpha-Heavy Chain) Ab-1
Immunohistochemistry (Paraffin)
Unlabeled
Murine
Immunoglobulins
Purified human alpha heavy chain
GA01
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa