missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ IgA (α -Heavy Chain) Ab-1, Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ IgA (α-Heavy Chain) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS1008PABX

 View more versions of this product

Catalog No. MS1008PABX


 

Specifications

Description

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ IgA (α-Heavy Chain) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: GA01

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Third constant domain (CH3) of the α-chain of IgA

Immunogen: Purified human α-heavy chain

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

IgA (alpha-Heavy Chain) Ab-1
Monoclonal
1mg/mL
Concentrated
Purified human alpha heavy chain
200μL
Immunoglobulins
Monoclonal
Immunohistochemistry (Paraffin)
GA01
Unlabeled
Murine
IgG2a kappa
RUO
Primary
Human