Please login to your online account to display your discounted pricing

Thermo Scientific IgA (α -Heavy Chain) Ab-1, Mouse Monoclonal Antibody


Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ IgA (α-Heavy Chain) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific  MS1008P1ABX

 View more versions of this product

Catalog No. MS1008P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen IgA (alpha-Heavy Chain) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GA01
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Immunogen Purified human alpha heavy chain
Isotype IgG2a kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline Immunoglobulins
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Recommended for Immunohistochemistry (Formalin/paraffin), Thermo Scientific™ IgA (α-Heavy Chain) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: GA01

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Third constant domain (CH3) of the α-chain of IgA

Immunogen: Purified human α-heavy chain

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO