Please login to your online account to display your discounted pricing

Ig lambda light chain Mouse anti-Human, APC, Clone: 1-155-2, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $390.05

Specifications

Antigen Ig lambda light chain
Clone 1-155-2
Host Species Mouse
Gene Alias Immunoglobulin L, IgL
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015370

 
life technologies
LS17999041
25 tests Each for $187.05

5015371

 
life technologies
LS17999042
100 tests Each for $390.05
Description & Specifications

Specifications

Antigen Ig lambda light chain
Clone 1-155-2
Host Species Mouse
Gene Alias Immunoglobulin L, IgL
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The monoclonal antibody 1-155-2 recognizes the lambda light chain of human immunoglobulin. Approximately 22-kDa, the Ig lambda chain is comprised of one variable and one constant region. Two lambda chains together with the heavy chain comprise the whole Ig structure. Expression of Ig lambda is typically found in less than half of B lymphocytes, and has been associated with neoplastic lymphomas.