Please login to your online account to display your discounted pricing

Ig kappa light chain Mouse anti-Human, PE, Clone: TB28-2, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $317.55

Specifications

Antigen Ig kappa light chain
Clone TB28-2
Host Species Mouse
Gene Alias Immunoglobulin, Ig k
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011330

 
affymetrix
12997041
25 tests Each for $143.55

5011331

 
affymetrix
12997042
100 tests Each for $317.55
Description & Specifications

Specifications

Antigen Ig kappa light chain
Clone TB28-2
Host Species Mouse
Gene Alias Immunoglobulin, Ig k
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The monoclonal antibody TB28-2 recognizes the kappa light chain of human immunoglobulin. Approximately 22-kDa, the Ig kappa chain is comprised of one variable and one constant region. Two kappa chains together with the heavy chain comprise the whole Ig structure. Expression of Ig kappa is typically found on more than half of B lymphocytes.