Please login to your online account to display your discounted pricing

anti-Mouse IFN alpha A Recombinant, Thermo Scientific™

Ms IFN alpha A Recombinant protein 121001 from Thermo Scientific™ specifically detects Ms IFN alpha A in Ms samples, and it is validated for ELISA, IP and WB

$496.65 - $496.65

Products
Catalog Number Mfr. No. Price Quantity    

PI121001

 
pierce biotechnology inc
121001
Each for $496.65
Description & Specifications

Ms IFN alpha A Recombinant protein 121001 from Thermo Scientific™ specifically detects Ms IFN alpha A in Ms samples, and it is validated for ELISA, IP and WB


Host Species: E. Coli
Cross Reactivity: Mouse
Formulation: >1x10^5 units purified Protein (Lot-specific) in PBS, with 0.1% BSA and no preservative;
Synonyms: Murine interferon Alpha, IFNA
Uniprot Accession: P01572
Gene Symbol: Ifna1
Entrez GeneID: 15962
MA5-16490 detects SLA CLASS II DR in Bovine and Porcine samples. MA5-16490 has been successfully used in Flow Cytometry procedures. MA5-16490 The immunogen is Porcine monocytes.