Please login to your online account to display your discounted pricing

IFN gamma Rat anti-Mouse, PerCP-Cy5.5, Clone: XMG1.2, eBioscience™

Rat Monoclonal Antibody

$145.00 - $442.25

Specifications

Antigen IFN gamma
Clone XMG1.2
Host Species Rat
Gene Alias Interferon-gamma, IFN-g, IFNg
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015878

 
life technologies
LS45731180
25μg Each for $145.00

501129625

 
life technologies
LS45731182
100μg Each for $442.25
Description & Specifications

Specifications

Antigen IFN gamma
Clone XMG1.2
Host Species Rat
Gene Alias Interferon-gamma, IFN-g, IFNg
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PerCP-Cy5.5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The XMG1.2 antibody reacts with mouse interferon (IFN) gamma. The XMG1.2 antibody is a neutralizing antibody. Mouse IFN gamma is a 20kDa factor produced by activated T, B and NK cells, and is an anti-viral and anti-parasitic cytokine. IFN gamma, in synergy with other cytokines such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity.