missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IFN gamma Rat anti-Mouse, eFluor 450, Clone: XMG1.2, eBioscience™

Rat Monoclonal Antibody

$172.55 - $382.80

Specifications

Antigen IFN gamma
Clone XMG1.2
Host Species Rat
Gene Alias Interferon-gamma, IFN-g, IFNg
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124215 Life Technologies
LS48731182
100μg Each for $382.80
5016399 Life Technologies
LS48731180
25μg Each for $172.55
Specifications

Description

The XMG1.2 antibody reacts with mouse interferon (IFN) gamma. The XMG1.2 antibody is a neutralizing antibody. Mouse IFN gamma is a 20kDa factor produced by activated T, B and NK cells, and is an anti-viral and anti-parasitic cytokine. IFN gamma, in synergy with other cytokines such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity.
Description & Specifications

Specifications

IFN gamma
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
XMG1.2
Interferon-gamma, IFN-g, IFNg
Flow Cytometry (Intracellular Staining)
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL