missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IFN gamma Rat anti-Mouse, Biotin, Clone: XMG1.2, eBioscience™

Rat Monoclonal Antibody

$130.68 - $341.88

Specifications

Antigen IFN gamma
Clone XMG1.2
Host Species Rat
Gene Alias Interferon-gamma, IFN-g, IFNg
Applications ELISA
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012195 Life Technologies
LS13731181
50μg Each for $130.68
5012196 Life Technologies
LS13731185
500μg Each for $341.88
Specifications

Description

The XMG1.2 antibody reacts with mouse interferon (IFN) gamma. The XMG1.2 antibody is a neutralizing antibody. Mouse IFN gamma is a 20kDa factor produced by activated T, B and NK cells, and is an anti-viral and anti-parasitic cytokine. IFN gamma, in synergy with other cytokines such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity.
Description & Specifications

Specifications

IFN gamma
Rat
ELISA
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
XMG1.2
Interferon-gamma, IFN-g, IFNg
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer