Please login to your online account to display your discounted pricing

IFN gamma Mouse anti-Human, PE-eFluor 610, Clone: 4S.B3, eBioscience™

Mouse Monoclonal Antibody

$205.90 - $487.20

Specifications

Antigen IFN gamma
Clone 4S.B3
Host Species Mouse
Gene Alias Interferon gamma
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124874

 
life technologies
LS61731941
25 tests Each for $205.90

501124875

 
life technologies
LS61731942
100 tests Each for $487.20
Description & Specifications

Specifications

Antigen IFN gamma
Clone 4S.B3
Host Species Mouse
Gene Alias Interferon gamma
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The 4S.B3 monoclonal antibody reacts with interferon-gamma (IFN gamma). Human IFN gamma is a 17kDa factor produced by activated T and NK cells and is an anti-viral and anti-parasitic cytokine. IFN gamma in synergy with other cytokines, such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity.