Please login to your online account to display your discounted pricing

IFN gamma Mouse anti-Human, FITC, Clone: 4S.B3, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $288.55

Specifications

Antigen IFN gamma
Clone 4S.B3
Host Species Mouse
Gene Alias Interferon gamma
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010030

 
life technologies
LS11731982
100μg Each for $288.55

501122160

 
life technologies
LS11731941
25 tests Each for $143.55

501122161

 
life technologies
LS11731981
50μg Each for $172.55
Description & Specifications

Specifications

Antigen IFN gamma
Clone 4S.B3
Host Species Mouse
Gene Alias Interferon gamma
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL; 5μL (0.5μg)/test

The 4S.B3 monoclonal antibody reacts with interferon-gamma (IFN gamma). Human IFN gamma is a 17kDa factor produced by activated T and NK cells and is an anti-viral and anti-parasitic cytokine. IFN gamma in synergy with other cytokines, such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity.