missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IFN gamma Mouse anti-Human, eFluor 660, Clone: 4S.B3, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $341.88

Specifications

Antigen IFN gamma
Clone 4S.B3
Host Species Murine
Gene Alias Interferon gamma
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124450 Life Technologies
LS50731941
25 Tests Each for $170.28
501124451 Life Technologies
LS50731942
100 Tests Each for $341.88
Specifications

Description

The 4S.B3 monoclonal antibody reacts with interferon-gamma (IFN gamma). Human IFN gamma is a 17kDa factor produced by activated T and NK cells and is an anti-viral and anti-parasitic cytokine. IFN gamma in synergy with other cytokines, such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity.
Description & Specifications

Specifications

IFN gamma
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
4S.B3
Interferon gamma
Flow Cytometry,Flow Cytometry (Intracellular Staining)
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test