missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IFN gamma Mouse anti-Human, Clone: 4S.B3, eBioscience™

Mouse Monoclonal Antibody

$104.28 - $302.28

Specifications

Antigen IFN gamma
Clone 4S.B3
Host Species Murine
Gene Alias Interferon gamma
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013686 Life Technologies
LS14731981
50μg Each for $104.28
5013687 Life Technologies
LS14731985
500μg Each for $302.28
Specifications

Description

The 4S.B3 monoclonal antibody reacts with interferon-gamma (IFN gamma). Human IFN gamma is a 17kDa factor produced by activated T and NK cells and is an anti-viral and anti-parasitic cytokine. IFN gamma in synergy with other cytokines, such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity.
Description & Specifications

Specifications

IFN gamma
Murine
Flow Cytometry (Intracellular Staining)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
4S.B3
Interferon gamma
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer