missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IFN gamma Mouse anti-Human, APC-eFluor 780, Clone: 4S.B3, eBioscience™

Mouse Monoclonal Antibody

$161.04 - $415.80

Specifications

Antigen IFN gamma
Clone 4S.B3
Host Species Murine
Gene Alias IFNg, Interferon gamma
Applications Flow Cytometry,Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245979 Life Technologies
LS47731941
25 Tests Each for $161.04
501128729 Life Technologies
LS47731942
100 Tests Each for $415.80
Specifications

Description

The 4S.B3 monoclonal antibody reacts with interferon-gamma (IFN gamma). Human IFN gamma is a 17kDa factor produced by activated T and NK cells and is an anti-viral and anti-parasitic cytokine. IFN gamma in synergy with other cytokines, such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity.
Description & Specifications

Specifications

IFN gamma
Murine
Flow Cytometry,Flow Cytometry (Intracellular Staining)
APC-eFluor 780
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.25μg)/test
4S.B3
IFNg, Interferon gamma
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer