Please login to your online account to display your discounted pricing

IFN gamma Mouse anti-Human, Alexa Fluor 488, Clone: 4S.B3, eBioscience™

Mouse Monoclonal Antibody

$162.40 - $442.25

Specifications

Antigen IFN gamma
Clone 4S.B3
Host Species Murine
Gene Alias Interferon-gamma
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016808

 
life technologies
LS53731941
25 Tests Each for $162.40

5016809

 
life technologies
LS53731942
100 Tests Each for $442.25
Description & Specifications

Specifications

Antigen IFN gamma
Clone 4S.B3
Host Species Murine
Gene Alias Interferon-gamma
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The 4S.B3 monoclonal antibody reacts with interferon-gamma (IFN gamma). Human IFN gamma is a 17kDa factor produced by activated T and NK cells and is an anti-viral and anti-parasitic cytokine. IFN gamma in synergy with other cytokines, such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity.