Learn More
IFN gamma Monoclonal Antibody (XMG1.2), Alexa Fluor™ 700, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$208.00 - $383.00
Specifications
Antigen | IFN gamma |
---|---|
Clone | XMG1.2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Description: The XMG1.2 antibody reacts with mouse interferon (IFN) gamma. The XMG1.2 antibody is a neutralizing antibody. Mouse IFN gamma is a 20 kDa factor produced by activated T, B and NK cells, and is an anti-viral and anti-parasitic cytokine. IFN gamma, in synergy with other cytokines such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN gamma. Glycosylation of IFN gamma does not affect its biological activity. Applications Reported: This XMG1.2 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This XMG1.2 antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.6 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Alexa Fluor™ 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome.
IFN gamma Monoclonal antibody specifically detects IFN gamma in Mouse samples. It is validated for Flow Cytometry.Specifications
IFN gamma | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN ?; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFN?; Immune interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon ?; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferon?; M-IFN-g; R-IFN-g | |
IFNG | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IFNG |
XMG1.2 | |
Flow Cytometry | |
Alexa Fluor 700 | |
Rat | |
Mouse | |
P01580 | |
15978 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |