missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IFN alpha receptor 1 (IFNAR1) Mouse anti-Mouse, PE, Clone: MAR1-5A3, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $341.88

Specifications

Antigen IFN alpha receptor 1 (IFNAR1)
Clone MAR1-5A3
Host Species Murine
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122358 Life Technologies
LS12594580
25μg Each for $157.08
501122359 Life Technologies
LS12594582
100μg Each for $341.88
Specifications

Description

The MAR1-5A3 monoclonal antibody reacts with mouse IFN alpha/beta receptor subunit 1 (IFNAR-1). The type I IFN receptor subunit 1 is co-expressed with IFNAR-2 on nearly all cells and make up the heterodimeric receptor complex that binds to all type I IFNs, IFN alpha and IFN beta. Type I IFNs are a group of structurally and functionally related proteins that demonstrate antiviral, antiparasitic, and antiproliferative activities.
Description & Specifications

Specifications

IFN alpha receptor 1 (IFNAR1)
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
MAR1-5A3
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer