Please login to your online account to display your discounted pricing

IFN alpha receptor 1 (IFNAR1) Mouse anti-Mouse, Clone: MAR1-5A3, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$2,898.55 - $2,898.55

Specifications

Antigen IFN alpha receptor 1 (IFNAR1)
Clone MAR1-5A3
Host Species Murine
Species Reactivity Mouse
Applications Cytokine Neutralization
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122971

 
life technologies
LS16594538
5mg Each for $2,898.55
Description & Specifications

Specifications

Antigen IFN alpha receptor 1 (IFNAR1)
Clone MAR1-5A3
Host Species Murine
Species Reactivity Mouse
Applications Cytokine Neutralization
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The MAR1-5A3 monoclonal antibody reacts with mouse IFN alpha/beta receptor subunit 1 (IFNAR-1). The type I IFN receptor subunit 1 is co-expressed with IFNAR-2 on nearly all cells and make up the heterodimeric receptor complex that binds to all type I IFNs, IFN alpha and IFN beta. Type I IFNs are a group of structurally and functionally related proteins that demonstrate antiviral, antiparasitic, and antiproliferative activities.