Please login to your online account to display your discounted pricing

Ifenprodil hemitartrate, Tocris


Non-competitive NMDA antagonist; Also σ ligand

Quantity 50mg

Manufacturer: r & d systems  RDS054550

 View more versions of this product

Catalog No. 054550

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 400.49
Chemical Name or Material Ifenprodil hemitartrate
Assay Percent Range >99%
Molecular Formula C21H27NO2·1/2C4H6O6
CAS 23210-56-2
Synonym (1R™, 2S™)-erythro-2-(4-Benzylpiperidino)-1-(4-hydroxyphenyl)-1-propanol hemitartrate
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Ifenprodil hemitartrate
Assay Percent Range >99%
Molecular Formula C21H27NO2·1/2C4H6O6
Formula Weight 400.49
Synonym (1R™, 2S™)-erythro-2-(4-Benzylpiperidino)-1-(4-hydroxyphenyl)-1-propanol hemitartrate
Accessories

Store at Room Temperature