Please login to your online account to display your discounted pricing

Ifenprodil hemitartrate, Tocris

Non-competitive NMDA antagonist; Also σ ligand

$113.57 - $437.93

Chemical Identifiers

Chemical Name or Material Ifenprodil hemitartrate
Assay Percent Range >99%
Molecular Formula C21H27NO2·1/2C4H6O6
Formula Weight 400.49
Synonym (1R™, 2S™)-erythro-2-(4-Benzylpiperidino)-1-(4-hydroxyphenyl)-1-propanol hemitartrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

054510

 
r & d systems
RDS054510
10mg Each for $113.57

054550

 
r & d systems
RDS054550
50mg Each for $437.93
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 400.49
Chemical Name or Material Ifenprodil hemitartrate
Assay Percent Range >99%
Molecular Formula C21H27NO2·1/2C4H6O6
CAS 23210-56-2
Synonym (1R™, 2S™)-erythro-2-(4-Benzylpiperidino)-1-(4-hydroxyphenyl)-1-propanol hemitartrate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature