Please login to your online account to display your discounted pricing

IEM 1754 dihydrobromide, Tocris™

Voltage-dependent, open-channel blocker of AMPA currents

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material IEM 1754 dihydrobromide
Assay Percent Range >98%
Molecular Formula C16H30N2·2HBr
Formula Weight 412.25
Synonym N-(Tricyclo[3.3.1.13, 7]dec-1-ylmethyl)-1, 5-pentanediamine dihydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

419910

 
r & d systems
RDS419910
10mg Each for $201.36

419950

 
r & d systems
RDS419950
50mg Each for $844.73
Description & Specifications

Specifications

Chemical Name or Material IEM 1754 dihydrobromide
Quantity 10mg
CAS 162831-31-4
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C16H30N2·2HBr
Formula Weight 412.25
Synonym N-(Tricyclo[3.3.1.13, 7]dec-1-ylmethyl)-1, 5-pentanediamine dihydrobromide
Safety and Handling

Store at Room Temperature