Please login to your online account to display your discounted pricing

IEM 1460, Tocris

Open-channel blocker of AMPA currents; selective for non-GluR2-containing receptors

$139.08 - $585.69

Chemical Identifiers

Chemical Name or Material IEM 1460
Assay Percent Range >98%
Molecular Formula C19H37N2Br·HBr
Formula Weight 454.33
Synonym N,N,H,-Trimethyl-5-[(tricyclo[3.3.1.13,7]dec-1-ylmethyl)amino]-1-pentanaminiumbromide hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

163610

 
r & d systems
RDS163610
10mg Each for $139.08

163650

 
r & d systems
RDS163650
50mg Each for $585.69
Description & Specifications

Specifications

Formula Weight 454.33
Quantity 10mg
Assay Percent Range >98%
CAS 121034-89-7
Chemical Name or Material IEM 1460
Molecular Formula C19H37N2Br·HBr
Synonym N,N,H,-Trimethyl-5-[(tricyclo[3.3.1.13,7]dec-1-ylmethyl)amino]-1-pentanaminiumbromide hydrobromide
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature