Please login to your online account to display your discounted pricing

Tocris™ IDRA 21


Inhibits AMPA receptor desensitization

Manufacturer: tocris™  121950

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 121950

  • / Each
 

Description & Specifications

Specifications

Purity >98
Inhibitors IDRA 21
Molecular Weight 232.68
Quantity 50mg
Product Type IDRA 21

Inhibits AMPA receptor desensitization and enhances cognition by a related mechanism. More able to cross the blood-brain barrier than cyclothiazide (Cat. No. 0713).

  • Chemical Name: 7-Chloro-3-methyl-3,4-dihydro-2H-1,2,4-benzothiadiazine S,S-dioxide
  • Formula: C8H9ClN2O2S
  • CAS Number: 22503-72-6
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >98