missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ IDRA 21

Inhibits AMPA receptor desensitization

$199.90 - $838.64

Specifications

Product Type IDRA 21
Inhibitors IDRA 21
Molecular Weight 232.68
Purity >98
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
121910 Tocris™
121910
10mg Each for $199.90
121950 Tocris™
121950
50mg Each for $838.64
Specifications

Description

Inhibits AMPA receptor desensitization and enhances cognition by a related mechanism. More able to cross the blood-brain barrier than cyclothiazide (Cat. No. 0713).
  • Chemical Name: 7-Chloro-3-methyl-3,4-dihydro-2H-1,2,4-benzothiadiazine S,S-dioxide
  • Formula: C8H9ClN2O2S
  • CAS Number: 22503-72-6
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >98
Description & Specifications

Specifications

IDRA 21
IDRA 21
232.68
>98