missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IDO Rat anti-Mouse, PerCP-eFluor 710, Clone: mIDO-48, eBioscience™

Rat Monoclonal Antibody

$223.08 - $526.68

Specifications

Antigen IDO
Clone mIDO-48
Host Species Rat
Gene Alias Indoleamine 2,3-dioxygenase 1
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123947 Life Technologies
LS46947380
25μg Each for $223.08
501128592 Life Technologies
LS46947382
100μg Each for $526.68
Specifications

Description

The monoclonal antibody mIDO-48 reacts with mouse indoleamine 2,3-dioxygenase 1(IDO1). IDO1 is an enzyme expressed intracellularly by dendritic cells, interferon gamma activated macrophages, epithelial cells, vascular endothelium and tumor cells. This enzyme catalyzes tryptophan into kynurenine metabolites. The reduction of tryptophan and the introduction of kynurenine metabolites in the microenvironment of IDO-expressing cells suppress T cell proliferation and promote the activation and induction of regulatory T cells.
Description & Specifications

Specifications

IDO
Rat
Flow Cytometry (Intracellular Staining)
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL
mIDO-48
Indoleamine 2,3-dioxygenase 1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer