missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IDO Monoclonal Antibody (mIDO-48), eFluor™ 660, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Supplier:  Invitrogen 50947382

 View more versions of this product

Catalog No. 501128978



Description

Description

Description: The monoclonal antibody mIDO-48 reacts with mouse indoleamine 2,3-dioxygenase 1(IDO1). IDO1 is an enzyme expressed intracellularly by dendritic cells, interferon gamma activated macrophages, epithelial cells, vascular endothelium and tumor cells. This enzyme catalyzes tryptophan into kynurenine metabolites. The reduction of tryptophan and the introduction of kynurenine metabolites in the microenvironment of IDO-expressing cells suppress T cell proliferation and promote the activation and induction of regulatory T cells. Applications Reported: This mIDO-48 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This mIDO-48 antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

IDO Monoclonal antibody specifically detects IDO in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

IDO
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P28776
Ido1
Affinity chromatography
RUO
15930
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
mIDO-48
eFluor 660
Ido1
EC 1.13.11.52; IDO; IDO1; IDO-1; INDO; indolamine 2,3 dioxygenase; indole 2,3-dioxygenase; indoleamine; indoleamine 2,3-dioxygenase 1; indoleamine 23-dioxygenase; Indoleamine-2; indoleamine-pyrrole 2,3 dioxygenase; indoleamine-pyrrole 2,3-dioxygenase
Rat
100 μg
Primary
Mouse
Antibody
IgG2b κ
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only.