Please login to your online account to display your discounted pricing

IDO Mouse anti-Human, PE-Cy7, Clone: eyedio, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $433.55

Specifications

Antigen IDO
Clone eyedio
Host Species Mouse
Gene Alias Indoleamine-2,3-dioxygenase
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123453

 
life technologies
LS25947741
25 tests Each for $187.05

501123454

 
life technologies
LS25947742
100 tests Each for $433.55
Description & Specifications

Specifications

Antigen IDO
Clone eyedio
Host Species Mouse
Gene Alias Indoleamine-2,3-dioxygenase
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

This eyedio monoclonal antibody reacts with human indoleamine-2,3-dioxygenase (IDO, INDO, IDO1), an intracellular enzyme that catalyzes the degradation of tryptophan to kynurenines. IDO is expressed in a wide variety of tissues and cells, including macrophages, plasmacytoid dendritic cells, and several cell lines. IDO can be induced in many different cell types by IFN gamma or other inflammatory stimuli. Expression of IDO in antigen presenting cells and tumors is thought to mediate immune suppression through depletion of this essential amino acid and/or by the creation of tryptophan metabolites that cause apoptosis of T cells and induction of regulatory T cells.