Please login to your online account to display your discounted pricing

IDO Mouse anti-Human, eFluor 660, Clone: V1NC3IDO, eBioscience™

Mouse Monoclonal Antibody

$146.45 - $371.20

Specifications

Antigen IDO
Clone V1NC3IDO
Host Species Murine
Gene Alias Indoleamine-2,3-dioxygenase
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124525

 
life technologies
LS50975080
25μg Each for $146.45

501124526

 
life technologies
LS50975082
100μg Each for $371.20
Description & Specifications

Specifications

Antigen IDO
Clone V1NC3IDO
Host Species Murine
Gene Alias Indoleamine-2,3-dioxygenase
Species Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody V1NC3IDO recognizes human IDO. IDO (indoleamine-2,3-dioxygenase, INDO, IDO1) is an intracellular enzyme that catalyzes the degradation of tryptophan to kynurenines. IDO is expressed in a wide variety of tissues and cells, including macrophages, plasmacytoid dendritic cells, and several cell lines. IDO can be induced in many different cell types by IFN gamma or other inflammatory stimuli. Expression of IDO in antigen presenting cells and tumors is thought to mediate immune suppression through depletion of this essential amino acid and/or by the creation of tryptophan metabolites that cause apoptosis of T cells and induction of regulatory T cells.

This V1NC3IDO antibody reacts with IDO1 and does not cross react with IDO2.