Idex Health & Science™ Very High Pressure (VHP) Filters DFS Item
Filters with inline and precolumn designs for UHPLC
$729.54 - $1,665.89
Specifications
For Use With (Application) Very high pressure
View More Specs
Products 4
Catalog Number Mfr. No. Pore Size Length (Metric) Product Type Price Quantity  
Catalog Number Mfr. No. Pore Size Length (Metric) Product Type Price Quantity  
05-701-470 View Documents Idex Health & Science™
UPVHP500
0.5μm 23.9mm Inline filter Each for $729.54
05-701-475 View Documents Idex Health & Science™
UPVHP555
0.2μm 34.8mm Precolumn filter Each for $1,665.89
05-701-474 View Documents Idex Health & Science™
UPVHP550
0.5μm 34.8mm Precolumn filter Each for $1,302.93
05-701-471 View Documents Idex Health & Science™
UPVHP505
0.2μm 23.9mm Inline filter Each for $760.26
Description
Description
  • Protect chromatography columns and other system components from harmful particulates
  • Captures particles from fluid stream before they damage expensive fluidic elements
  • Designed to withstand 20,000+psi (1380+ bar)
  • Available pore sizes: 0.2μm and 0.5μm
  • Each assembly comes with four spare filter units
Inline filters
  • Placed between high pressure pump and injection valve in UHPLC system
  • Catch particles stemming from pump seal wear
  • Small internal volume of these filters allows gradient changes to reach column with minimal delay
Precolumn filters
  • Connect directly to 10-32 coned port of chromatography column
  • Adjustable tube tip for various port depths, preventing formation of dead volume
  • Reusable in multiple ports, with ensured good connections
Specifications
Specifications
Very high pressure