Idex Health & Science™ Very High Pressure (VHP) Filters DFS Item
Filters with inline and precolumn designs for UHPLC
Manufacturer:  Idex Health & Science™ UPVHP555
 View more versions of this product
Catalog No. 05-701-475
Description
Description
  • Protect chromatography columns and other system components from harmful particulates
  • Captures particles from fluid stream before they damage expensive fluidic elements
  • Designed to withstand 20,000+psi (1380+ bar)
  • Available pore sizes: 0.2μm and 0.5μm
  • Each assembly comes with four spare filter units
Inline filters
  • Placed between high pressure pump and injection valve in UHPLC system
  • Catch particles stemming from pump seal wear
  • Small internal volume of these filters allows gradient changes to reach column with minimal delay
Precolumn filters
  • Connect directly to 10-32 coned port of chromatography column
  • Adjustable tube tip for various port depths, preventing formation of dead volume
  • Reusable in multiple ports, with ensured good connections
Specifications
Specifications
0.2μm
Very high pressure
34.8mm
Proprietary PEEK™ Polymer Blend/316 Stainless Steel
1.37 in.
Precolumn filter
Documents