Please login to your online account to display your discounted pricing

Idebenone, Tocris

Antioxidant and neuroprotective agent

$95.70 - $375.15

Chemical Identifiers

Chemical Name or Material Idebenone
Assay Percent Range >99%
Molecular Formula C19H30O5
Formula Weight 338.44
Synonym 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

300250

 
r & d systems
RDS300250
50mg Each for $375.15

300210

 
r & d systems
RDS300210
10mg Each for $95.70
Description & Specifications

Specifications

Formula Weight 338.44
Quantity 50mg
Synonym 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Chemical Name or Material Idebenone
Molecular Formula C19H30O5
CAS 58186-27-9
Safety and Handling

Store at Room Temperature