Please login to your online account to display your discounted pricing

IDE 2, Tocris™

Induces definitive endoderm formation in mouse and human ESCs

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material IDE 2
Assay Percent Range >98%
Molecular Formula C12H20N2O3
Formula Weight 240.3
Synonym Heptanedioic acid-1-(2-cyclopentylidenehydrazide)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

401650

 
r & d systems
RDS401650
50mg Each for $924.23

401610

 
r & d systems
RDS401610
10mg Each for $225.77
Description & Specifications

Specifications

Formula Weight 240.3
Quantity 50mg
Chemical Name or Material IDE 2
Assay Percent Range >98%
Molecular Formula C12H20N2O3
CAS 1136466-93-7
Synonym Heptanedioic acid-1-(2-cyclopentylidenehydrazide)
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C