Please login to your online account to display your discounted pricing

ID 8, Tocris™

Sustains self-renewal and pluripotency of ESCs

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material ID 8
Assay Percent Range >99%
Molecular Formula C16H14N2O4
Formula Weight 298.29
Synonym 1-(4-Methoxyphenyl)-2-methyl-3-nitro-1H-indol-6-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

385310

 
r & d systems
385310
10mg Each for $221.01

385350

 
r & d systems
385350
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 298.29
Quantity 10mg
Chemical Name or Material ID 8
Molecular Formula C16H14N2O4
Assay Percent Range >99%
CAS 147591-46-6
Synonym 1-(4-Methoxyphenyl)-2-methyl-3-nitro-1H-indol-6-ol
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C