Please login to your online account to display your discounted pricing

Alfa Aesar™ ICP-MS Stock Standard solution B for 200.8, Specpure™

Chemical Identifiers

Chemical Name or Material ICP-MS Stock Standard solution B for 200.8
Purity Grade Specpure™
MDL Number MFCD00803327
UN Number UN3264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40486AE

  SDS
alfa aesar™
40486AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 2 GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 100mL
Concentration Matrix: 1% HNO3
TSCA TSCA
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
MDL Number MFCD00803327
Purity Grade Specpure™
Physical Form Liquid
UN Number UN3264
Chemical Name or Material ICP-MS Stock Standard solution B for 200.8
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.