Please login to your online account to display your discounted pricing

Alfa Aesar™ ICP-MS Memory Check Standard solution B for 6020 CLP-M, Specpure™

Chemical Identifiers

Chemical Name or Material ICP-MS Memory Check Standard solution B for 6020 CLP-M
Purity Grade Specpure™
MDL Number MFCD00803323
UN Number UN3264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40488AE

  SDS
alfa aesar™
40488AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40488AP

  SDS
alfa aesar™
40488AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 100mL
TSCA TSCA
UN Number UN3264
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Density 1.0g/mL
Concentration Matrix: 2% HNO3/tr. F-/tr. tartaric acid
Physical Form Liquid
Chemical Name or Material ICP-MS Memory Check Standard solution B for 6020 CLP-M
MDL Number MFCD00803323
Health Hazard 1 Warning
Purity Grade Specpure™
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.