Please login to your online account to display your discounted pricing

Alfa Aesar™ ICP-MS Interferents A for 6020A ICS, Specpure™

Chemical Identifiers

Chemical Name or Material ICP-MS Interferents A for 6020A ICS
Purity Grade Specpure™
MDL Number MFCD08064524
UN Number UN3264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47135AP

  SDS
alfa aesar™
47135AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

UN Number UN3264
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Quantity 500mL
MDL Number MFCD08064524
Avg. Mol. Wt. or Mol. Wt. Range Matrix: 2% HNO3/6.5% HCl/tr· HFC5
TSCA TSCA
Physical Form Liquid
Health Hazard 1 Warning
Purity Grade Specpure™
Chemical Name or Material ICP-MS Interferents A for 6020A ICS
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.