missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ ICP-MS General Memory& Interference Check Sample Mix for 6020, Specpure™

Manufacturer:  Alfa Aesar™ 44935AP

 View more versions of this product

Catalog No. AA44935AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

ICP-MS General Memory & Interference Check Sample Mix for 6020
Liquid
500mL
UN3264
Specpure™
Matrix: 2% HNO3
Documents