missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ICP-MS Calibration Standard 1, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,7697-37-2,7784-27-2,7789-02-8,7647-01-0,13823-29-5
CAS Min % 89.0,7.23,0.01,0.01,0.01,0.0023
CAS Max % 92.0,7.53,0.01,0.01,0.01,0.0023
Chemical Name or Material ICP-MS Calibration Standard 1
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
RICPMS1-100 Ricca Chemical Company
RIRICPMS1100
Poly natural 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
RMSCAL1A-100 Ricca Chemical Company
RIRMSCAL1A100
Poly natural 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7697-37-2,7784-27-2,7789-02-8,7647-01-0,13823-29-5
89.0,7.23,0.01,0.01,0.01,0.0023
92.0,7.53,0.01,0.01,0.01,0.0023
ICP-MS Calibration Standard 1

Specifications

100mL
Clear colorless
92.0,0.01,0.01,7.53,0.0023,0.01
Poly natural
Odorless
7732-18-5,7697-37-2,13823-29-5,7789-02-8,7647-01-0,7784-27-2
7.23,89.0,0.01,0.0023,0.01,0.01
ICP-MS Calibration Standard 1

Safety and Handling

ShelfLife : 12 months