Please login to your online account to display your discounted pricing

Alfa Aesar™ Icilin, 97%


Quantity 10mg

Manufacturer: alfa aesar™  J67451MA

 View more versions of this product

Catalog No. AAJ67451MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 311.3
Chemical Name or Material Icilin
MDL Number MFCD01689072
CAS 36945-98-9
Assay 97%
Synonym AG 3-5; 3,6-Dihydro-1-(2-hydroxyphenyl)-4-(3-nitrophenyl)-2(1H)-pyrimidinone
Color Yellow to orange
Quantity 10mg
RTECS Number UW7397500
Physical Form Solid

Specifications

Chemical Name or Material Icilin
CAS 36945-98-9
Formula Weight 311.3
MDL Number MFCD01689072
Synonym AG 3-5; 3,6-Dihydro-1-(2-hydroxyphenyl)-4-(3-nitrophenyl)-2(1H)-pyrimidinone