Please login to your online account to display your discounted pricing

Alfa Aesar™ Icilin, 97%

Chemical Identifiers

Chemical Name or Material Icilin
CAS 36945-98-9
Formula Weight 311.3
MDL Number MFCD01689072
Synonym AG 3-5; 3,6-Dihydro-1-(2-hydroxyphenyl)-4-(3-nitrophenyl)-2(1H)-pyrimidinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67451MA

 
alfa aesar™
J67451MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 311.3
Chemical Name or Material Icilin
MDL Number MFCD01689072
CAS 36945-98-9
Assay 97%
Synonym AG 3-5; 3,6-Dihydro-1-(2-hydroxyphenyl)-4-(3-nitrophenyl)-2(1H)-pyrimidinone
Color Yellow to orange
Quantity 10mg
RTECS Number UW7397500
Physical Form Solid