Please login to your online account to display your discounted pricing

ICI 63197, Tocris

PDE4 inhibitor

$175.83 - $734.99

Chemical Identifiers

Chemical Name or Material ICI 63197
Assay Percent Range >99%
Molecular Formula C9H13N5O
Formula Weight 207.23
Synonym 2-Amino-6-methyl-4-propyl-[1,2,4]triazolo[1,5-a]pyrimidin-5(4H)-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

181650

 
r & d systems
181650
50mg Each for $734.99

181610

 
r & d systems
181610
10mg Each for $175.83
Description & Specifications

Specifications

Formula Weight 207.23
Quantity 50mg
Chemical Name or Material ICI 63197
Assay Percent Range >99%
Molecular Formula C9H13N5O
CAS 27277-00-5
Synonym 2-Amino-6-methyl-4-propyl-[1,2,4]triazolo[1,5-a]pyrimidin-5(4H)-one
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C