Please login to your online account to display your discounted pricing

ICI 215,001 hydrochloride, Tocris


β3 agonist

Quantity 10mg

Manufacturer: r & d systems  RDS092910

 View more versions of this product

Catalog No. 092910

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 397.86
Synonym (S)-4-[2-Hydroxy-3-phenoxypropylaminoethoxy]phenoxyacetic acid hydrochloride
Chemical Name or Material ICI 215, 001 hydrochloride
CAS 140850-02-8
Molecular Formula C19H23NO6·HCl
Assay Percent Range >98%
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material ICI 215, 001 hydrochloride
Assay Percent Range >98%
Molecular Formula C19H23NO6·HCl
Formula Weight 397.86
Synonym (S)-4-[2-Hydroxy-3-phenoxypropylaminoethoxy]phenoxyacetic acid hydrochloride
Accessories

Store at Room Temperature